Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR161 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533654
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533654 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533654 Supplier Invitrogen™ Supplier No. PA533654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Elephant, Panda, Dog, Bovine, Horse, Rabbit, Pig (100%) Mouse, Opossum (95%).

Upon ligand binding, G protein-coupled receptors, such as GPR161, activate cytoplasmic G proteins (see GNAS, MIM 139320), allowing the receptors to transduce extracellular signals across the plasma membrane into the cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR161
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR161
Gene Accession No. Q2YDN1, Q8N6U8
Gene Alias FLJ33952; G protein-coupled receptor 161; Gm208; Gm208Gpr; Gpr161; G-protein coupled receptor 161; G-protein coupled receptor RE2; RE2; RGD1563245; Unknown (protein for MGC:134300); vl
Gene Symbols GPR161
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from C-terminus of human GPR161.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100051574, 100155594, 100349764, 23432, 490358, 768006
Target Species Human, Canine, Rabbit, Monkey, Bovine, Pig, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.