Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR161 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111687
Description
Immunogen sequence: NKTVRKELLG MCFGDRYYRE PFVQRQRTSR LFSISNRITD LGLSPHLTAL MAGGQPLGHS SSTGDTGFSC SQDSGTDM.
Upon ligand binding, G protein-coupled receptors, such as GPR161, activate cytoplasmic G proteins (see GNAS, MIM 139320), allowing the receptors to transduce extracellular signals across the plasma membrane into the cell.
Specifications
GPR161 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N6U8 | |
GPR161 | |
Recombinant Protein corresonding to Human GPR161. Recombinant protein control fragment (Product #RP-107859). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry | |
Unconjugated | |
GPR161 | |
FLJ33952; G protein-coupled receptor 161; Gm208; Gm208Gpr; Gpr161; G-protein coupled receptor 161; G-protein coupled receptor RE2; RE2; RGD1563245; Unknown (protein for MGC:134300); vl | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23432 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction