Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR162 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114738
Description
Antibody detects endogenous levels of total GPR162.
This gene was identified upon genomic analysis of a gene-dense region at human chromosome 12p13. It appears to be mainly expressed in the brain; however, its function is not known. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
GPR162 | |
Polyclonal | |
Unconjugated | |
GPR162 | |
A-2; BC031437; G protein-coupled receptor 162; gene rich cluster, A; gene-rich cluster gene A protein; GPR162; GRCA; probable G-protein coupled receptor 162 | |
Rabbit | |
Affinity chromatography | |
RUO | |
14788, 27239 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q16538, Q3UN16 | |
GPR162 | |
A synthesized peptide derived from human GPR162(Accession Q16538), corresponding to amino acid residues F54-T104. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction