Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR162 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983722
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3722 Supplier Bioss Supplier No. BS15386R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene was identified upon genomic analysis of a gene-dense region at human chromosome 12p13. It appears to be mainly expressed in the brain; however, its function is not known. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Antigen GPR162
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GPR162
Gene Accession No. Q16538
Gene Alias A-2; BC031437; BOS_5975; G protein-coupled receptor 162; gene rich cluster, A; Gene-rich cluster gene A protein; GPR162; GRCA; Probable G-protein coupled receptor 162; prolyl 3-hydroxylase 3
Gene Symbols GPR162
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPR162.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27239
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.