Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR164 Antibody, Novus Biologicals™
SDP

Catalog No. p-200047233 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB438340 25ul
NBP213701 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB438340 Supplier Novus Biologicals Supplier No. NBP21370125UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

GPR164 Polyclonal specifically detects GPR164 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GPR164
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias D-GPCR, Dresden-G-protein-coupled receptor, FLJ13581, GPR136, GPR164, G-protein coupled receptor 164, MGC24137, olfactory receptor 51E1, Olfactory receptor 52A3, olfactory receptor OR11-15, olfactory receptor, family 51, subfamily E, member 1, olfactory receptor, family 51, subfamily E, member 1 pseudogene, olfactory receptor, family 52, subfamily A, member 3 pseudogene, OR51E1P, OR52A3P, POGR, prostate overexpressed G protein-coupled receptor, Prostate-overexpressed G protein-coupled receptor, Prostate-specific G protein-coupled receptor 2, PSGR2
Gene Symbols OR51E1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: VKTKEIRQRILRLFHVATHASEP
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 143503
Test Specificity Specificity of human GPR164 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.