Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR164 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095295 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095295 20 μL
NB095294 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095295 Supplier Novus Biologicals Supplier No. NBP30297220UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR164 Polyclonal specifically detects GPR164 in Human samples. It is validated for Western Blot.

Specifications

Antigen GPR164
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias D-GPCR, Dresden-G-protein-coupled receptor, FLJ13581, GPR136, GPR164, G-protein coupled receptor 164, MGC24137, olfactory receptor 51E1, Olfactory receptor 52A3, olfactory receptor OR11-15, olfactory receptor, family 51, subfamily E, member 1, olfactory receptor, family 51, subfamily E, member 1 pseudogene, olfactory receptor, family 52, subfamily A, member 3 pseudogene, OR51E1P, OR52A3P, POGR, prostate overexpressed G protein-coupled receptor, Prostate-overexpressed G protein-coupled receptor, Prostate-specific G protein-coupled receptor 2, PSGR2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human GPR164 (NP_689643.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 143503
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.