Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR17 Mouse anti-Human, Clone: OTI10B8, lyophilized, TrueMAB™
SDP

Catalog No. 501670822
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-0822 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-0822 Supplier OriGene Supplier No. CF811748
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR17 is an orphan G protein-coupled receptor involved in orchestration of oligodendrocyte differentiation and myelination in the central nervous system. GPR17 is expressed in brain, kidney, heart and umbilical vein endothelial cells. GPR17 has dual specificity for uracil nucleotides and cysteinyl leukotrienes and signals through G(i) and inhibition of adenylyl cyclase. May mediate brain damage by nucleotides and cysteinyl leukotrienes following ischemia.

Specifications

Antigen GPR17
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI10B8
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene GPR17
Gene Accession No. Q13304
Gene Alias G protein-coupled receptor 17
Gene Symbols GPR17
Host Species Mouse
Immunogen Synthetic peptide (the amino acid sequence is considered to be commercially sensitive) within Human GPR17 C-terminal. The exact sequence is proprietary.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2840
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.