Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533659
Description
This antibody is not recommended for use in ICC applications. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Elephant, Panda, Bovine, Horse, Rabbit, Pig (100%).
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR17 is an orphan G protein-coupled receptor involved in orchestration of oligodendrocyte differentiation and myelination in the central nervous system. GPR17 is expressed in brain, kidney, heart and umbilical vein endothelial cells. GPR17 has dual specificity for uracil nucleotides and cysteinyl leukotrienes and signals through G(i) and inhibition of adenylyl cyclase. May mediate brain damage by nucleotides and cysteinyl leukotrienes following ischemia.
Specifications
GPR17 | |
Polyclonal | |
Unconjugated | |
GPR17 | |
AI853548; DKFZp686M18273; G protein-coupled receptor 17; Gpr17; gpr17 {ECO:0000312; G-protein coupled receptor 17; P2Y-like receptor; R12; RGD:1589785}; UDP/CysLT receptor; Uracil nucleotide/cysteinyl leukotriene receptor; Y12546; Z94155 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100357734, 102148121, 102162410, 2840, 782442 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q13304 | |
GPR17 | |
Synthetic 15 amino acid peptide from 1st cytoplasmic domain of human GPR17. | |
50 μg | |
Primary | |
Human, Rabbit, Monkey, Bovine, Pig, Horse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction