Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR171 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP314351
This item is not returnable.
View return policy
Description
GPR171 Polyclonal antibody specifically detects GPR171 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
GPR171 | |
Polyclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Antibody |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
F730001G15Rik, G protein-coupled receptor 171, G-protein coupled receptor H963, H963platelet activating receptor homolog, probable G-protein coupled receptor 171 | |
Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human GPR171. | |
50 μg | |
GPCR | |
29909 | |
Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction