Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR171 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532807
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Bovine, Rabbit (100%) Mouse, Rat, Dog, Bat, Elephant, Panda (94%).
GPR171 is a protein coding gene. Orphan receptor.
Specifications
GPR171 | |
Polyclonal | |
Unconjugated | |
GPR171 | |
A630006E05; F730001G15Rik; G protein-coupled receptor 171; Gpr171; G-protein coupled receptor H963; H963; platelet activating receptor homolog; probable G-protein coupled receptor 171 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
103350416, 29909, 767929 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O14626, Q3ZBK9 | |
GPR171 | |
Synthetic 16 amino acid peptide from 3rd extracellular domain of human GPR171. | |
50 μg | |
Primary | |
Human, Rabbit, Monkey, Bovine | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction