Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR171 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987336
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7336 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7336 Supplier Bioss Supplier No. BS6564R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR171 is a protein coding gene. Orphan receptor.

Specifications

Antigen GPR171
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GPR171
Gene Accession No. O14626
Gene Alias A630006E05; F730001G15Rik; G protein-coupled receptor 171; GPR171; G-protein coupled receptor H963; H963; platelet activating receptor homolog; Probable G-protein coupled receptor 171
Gene Symbols GPR171
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPR171.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29909
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.