Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR171 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303890100UL
Description
GPR171 Polyclonal specifically detects GPR171 in Rat samples. It is validated for Western Blot.Specifications
GPR171 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Rat | |
Purified |
Western Blot | |
Unconjugated | |
F730001G15Rik, G protein-coupled receptor 171, G-protein coupled receptor H963, H963platelet activating receptor homolog, probable G-protein coupled receptor 171 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human GPR171 (NP_037440.3). | |
100 μL | |
GPCR, Phospho Specific | |
29909 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction