Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR173 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533664
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533664 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533664 Supplier Invitrogen™ Supplier No. PA533664
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Dog, Bat, Bovine, Rabbit (100%) Panda (95%) Hamster, Elephant (90%).

This gene encodes a member of the G-protein coupled receptor 1 family. This protein contains 7 transmembrane domains and conserved cysteine residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR173
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR173
Gene Accession No. Q5E9H8, Q6PI62, Q9JJH2, Q9NS66
Gene Alias 3230401K02Rik; G protein-coupled receptor 173; G protein-coupled receptor SREB3; GPR173; G-protein coupled receptor 173; Probable G-protein coupled receptor 173; RP1-290F12.1; SREB3; Super conserved receptor expressed in brain 3
Gene Symbols GPR173
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from C-terminus of human GPR173.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100341211, 491892, 515148, 54328, 64021, 70771
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.