Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR173 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120315
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120315 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120315 Supplier Invitrogen™ Supplier No. PA5120315
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HT-29, U-251MG, SKOV3, A-549, Mouse small intestine, Mouse testis. The target is usually found in the following locations: Cell membrane, Multi-pass membrane protein. Immunogen sequence: FDVGTYKFIR EEDQCIFEHR YFKANDTLGF MLMLAVLMAA THAVYGKLLL FEYRHRKMKP VQMVPAISQN W.

This gene encodes a member of the G-protein coupled receptor 1 family. This protein contains 7 transmembrane domains and conserved cysteine residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR173
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GPR173
Gene Accession No. Q6PI62, Q9NS66
Gene Alias 3230401K02Rik; G protein-coupled receptor 173; G protein-coupled receptor SREB3; GPR173; G-protein coupled receptor 173; Probable G-protein coupled receptor 173; RP1-290F12.1; SREB3; Super conserved receptor expressed in brain 3
Gene Symbols GPR173
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-230 of human GPR173 (NP_061842.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54328, 70771
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.