Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR174 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599007
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599007 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599007 Supplier Invitrogen™ Supplier No. PA599007
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR174 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR174 (G protein-coupled receptor 174), also known as FKSG79, is a 333 amino acid multi-pass membrane protein that belongs to the G-protein coupled receptor 1 family and is thought to function as a receptor for G protein-coupled purines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR174
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene GPR174
Gene Accession No. Q9BXC1
Gene Alias FKSG79; G protein-coupled receptor 174; Gm376; GPCR17; GPR174; Jeg18; LYPSR3; probable G-protein coupled receptor 174; putative purinergic receptor FKSG79; RGD1564689
Gene Symbols GPR174
Host Species Rabbit
Immunogen Recombinant Human Probable G-protein coupled receptor 174 protein (291-333AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84636
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.