Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR176 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3046 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3046 Supplier Invitrogen™ Supplier No. PA3046
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

GPR176 is in the G protein-coupled receptor family and are cell surface receptors involved in responses to hormones, growth factors, and neurotransmitters. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster, Elephant, Bovine, Dog, Bat, Horse, Stickleback, Zebrafish (100%). GPR176 exists in different glycosylated forms.

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR176 (G protein-coupled receptor 176), also known as HB-954, GPR or Gm1012, is a 515 amino acid multi-pass membrane protein belonging to the G-protein coupled receptor 1 family. Expressed in brain and spleen, with trace expression in kidney, GPR176 functions as an orphan receptor that is thought to play a role in signaling events throughout the cell. Containing four N-glycosylation sites, seven transmembrane domains and a large C-terminal cytosolic domain, GPR176 is encoded by a gene mapping to human chromosome 15q14.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR176
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole Serum with 0.05% sodium azide
Gene GPR176
Gene Accession No. Q14439, Q80WT4
Gene Alias Agr9; G protein-coupled receptor 176; Gm1012; Gpr; GPR176; G-protein coupled receptor 176; G-protein coupled receptor AGR9; HB954; HB-954; probable G-protein coupled receptor 176; putative G protein coupled receptor; RBU-15
Gene Symbols GPR176
Host Species Rabbit
Immunogen KLH conjugated COOH-terminal amino acids of 496-515 (RVERKMSRNNKVSIFPKVDS).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11245, 381413
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.