Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR177 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100549 Supplier Invitrogen™ Supplier No. PA5100549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR177.

Regulates Wnt proteins sorting and secretion in a feedback regulatory mechanism. This reciprocal interaction plays a key role in the regulation of expression, subcellular location, binding and organelle-specific association of Wnt proteins. Plays also an important role in establishment of the anterior-posterior body axis formation during development. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR177
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene WLS
Gene Accession No. Q5T9L3, Q6DID7, Q6P689
Gene Alias 5031439A09Rik; AI173978; AI987742; C1orf139; evenness interrupted homolog; EVI; G protein-coupled receptor 177; Gpr177; Integral membrane protein GPR177; mig-14; MRP; Protein evenness interrupted homolog; protein wntless homolog; Putative NF-kappa-B-activating protein 373; putative NFkB activating protein 373; UNQ85/PRO18667; WLS; Wnt ligand secretion mediator; wntless homolog; wntless homolog (Drosophila); wntless Wnt ligand secretion mediator
Gene Symbols WLS
Host Species Rabbit
Immunogen A synthesized peptide derived from human WLS(Accession Q5T9L3), corresponding to amino acid residues F473-T523.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362065, 68151, 79971
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.