Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR177 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567054 Supplier Invitrogen™ Supplier No. PA567054
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAYRDDAFAEW TEMAHERVPR KLKCTFTSPK TPEHEGRYYE CDVLPFMEIG SVAHKFYLLN IRLPVNEKKK INVGIGEIKD Highest antigen sequence identity to the following orthologs - mouse 94%, rat 93%.

Regulates Wnt proteins sorting and secretion in a feedback regulatory mechanism. This reciprocal interaction plays a key role in the regulation of expression, subcellular location, binding and organelle-specific association of Wnt proteins. Plays also an important role in establishment of the anterior-posterior body axis formation during development. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR177
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WLS
Gene Accession No. Q5T9L3
Gene Alias 5031439A09Rik; AI173978; AI987742; C1orf139; evenness interrupted homolog; EVI; G protein-coupled receptor 177; Gpr177; Integral membrane protein GPR177; mig-14; MRP; Protein evenness interrupted homolog; protein wntless homolog; Putative NF-kappa-B-activating protein 373; putative NFkB activating protein 373; UNQ85/PRO18667; WLS; Wnt ligand secretion mediator; wntless homolog; wntless homolog (Drosophila); wntless Wnt ligand secretion mediator
Gene Symbols WLS
Host Species Rabbit
Immunogen Recombinant Human GPR177. Recombinant protein control fragment (Product #RP-105477).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79971
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.