Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR179 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553657 Supplier Invitrogen™ Supplier No. PA553657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNMLLQANDI RESSVEEDVE WYQALVRSVA EGDPRVYRAL LTFNPPPGAS HLQLALQATR TGEETILQDL SGNWVQEENP PGDLDTPALK KRVLTNDLGS LGSPKWPQAD GYVGDTQQVR LSPPFLECQE GRLRPGWLIT LSATF Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.

Probable G-protein coupled receptor 179 is a protein that in humans is encoded by the GPR179 gene. This gene encodes a member of the glutamate receptor subfamily of G protein-coupled receptors. The encoded protein has an EGF-like calcium binding domain and a seven transmembrane domain in the N-terminal region of the protein. Mutations in this gene are associated with congenital stationary night blindness type 1E.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR179
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPR179
Gene Accession No. Q6PRD1
Gene Alias 5330439C02Rik; CSNB1E; G protein-coupled receptor 179; GPR158L; GPR158L1; GPR158-like; GPR158-like 1; GPR158-like 1 receptor; GPR179; Probable G-protein coupled receptor 158-like 1; probable G-protein coupled receptor 179
Gene Symbols GPR179
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR179. Recombinant protein control fragment (Product #RP-91290).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440435
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.