Learn More
Invitrogen™ GPR18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532810
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Bat, Elephant, Horse, Rabbit, Pufferfish (100%); Mouse, Rat, Bovine, Pig, Opossum, Salmon, Zebrafish, Stickleback (94%); Chicken, Platypus, Lizard, Catfish (88%); Hamster, Turkey (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR18 is an orphan G protein-coupled receptor found at high levels in the testis, small intestines and cells associated with the immune system including lymphocytes, thymus and spleen. GPR18 has been found to be a receptor for endogenous lipid neurotransmitters, several of which also bind to cannabinoid receptors. Some ligands described to be GPR18 agonists or partial agonists include N-Arachidonoylglycine, Abnormal cannabidiol, O-1602, AM-251, Cannabidiol, delta 9-Tetrahydrocannabinol, N-arachidonoyl ethanolamine and Arachidonylcyclopropylamide. Antagonists include the synthetic cannabidiol related compounds CID-85469571 and O-1918.
Specifications
GPR18 | |
Polyclonal | |
Unconjugated | |
GPR18 | |
EMBL:AAI28783.1}; G protein-coupled receptor 18; GPCRW; Gpr18; gpr18 {ECO:0000312; G-protein coupled receptor 18; NAGly receptor; N-arachidonyl glycine receptor; RP11-461N23.8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100147633, 100357321, 2841 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q14330 | |
GPR18 | |
Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human GPR18. | |
50 μg | |
Primary | |
Human, Rabbit, Monkey, Horse, Bat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.