Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR180 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561477
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561477 Supplier Invitrogen™ Supplier No. PA561477
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLSKAQLTMT MNQTEHNLTV SQIPSPQTWH VFYADKYTCQ DDKENSQVED IPFEMVLLNP DAEGNPFDHF SAGES Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 81%.

This gene encodes a protein that is a member of the G protein-coupled receptor superfamily. This protein is produced predominantly in vascular smooth muscle cells and may play an important role in the regulation of vascular remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR180
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPR180
Gene Accession No. Q86V85
Gene Alias AB041545; E130016I23Rik; G protein-coupled receptor 180; GPR180; Integral membrane protein GPR180; intimal thickness-related receptor; ITR; MNCb-3029
Gene Symbols GPR180
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR180. Recombinant protein control fragment (Product #RP-98496).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160897
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.