Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR182 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532811
Description
Percent identity with other species by BLAST analysis: Human (100%) Gorilla, Gibbon (94%).
Adrenomedullin (ADM) is a hypotensive peptide that belongs to a peptide superfamily, which includes the calcitonin gene-related peptide (CGRP) and amylin. ADM was originally identified in the adrenal medulla, where it is highly expressed. It is also produced by most contractile cells and is upregulated during sepsis and ischemia. Three distinct receptors have the ability to bind ADM and are designated ADM receptor (also designated L1), RDC-1 and the Calcitonin Receptor-Like Receptor (CRLR). The CRLR associates with receptor activity-modifying proteins (RAMPs), which determine the specificity of CRLR binding. Co-expression with RAMP1 results in CRLR binding to CGRP, whereas association with RAMP2 or 3 results in ADM binding. The ADM receptor, a seven transmembrane G protein-coupled receptor, specifically binds ADM and is highly expressed in heart, brain, skeletal muscle, the immune system, adrenal gland and liver. The ADM receptor mediates the signals produced by ADM through G proteins, which activate adenylate cyclase.
Specifications
GPR182 | |
Polyclonal | |
Unconjugated | |
GPR182 | |
7TMR; ADMR; adrenomedullin receptor; AMR; AM-R; G protein-coupled receptor 182; G10D; G10-D; gamrh; Gpcr17; Gpcr22; GPR182; G-protein coupled receptor 182; HAMR; hrhAMR; MB10; NOW; RAMR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11318 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
PBS with 0.1% sodium azide | |
O15218 | |
GPR182 | |
Synthetic 18 amino acid peptide from C-terminus of human GPR182 / ADMR. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction