Learn More
Invitrogen™ GPR182 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532814
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%); Elephant (94%); Marmoset, Bat, Panda, Rabbit, Horse (89%); Mouse, Rat, Hamster, Pig (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Adrenomedullin (ADM) is a hypotensive peptide that belongs to a peptide superfamily, which includes the calcitonin gene-related peptide (CGRP) and amylin. ADM was originally identified in the adrenal medulla, where it is highly expressed. It is also produced by most contractile cells and is upregulated during sepsis and ischemia. Three distinct receptors have the ability to bind ADM and are designated ADM receptor (also designated L1), RDC-1 and the Calcitonin Receptor-Like Receptor (CRLR). The CRLR associates with receptor activity-modifying proteins (RAMPs), which determine the specificity of CRLR binding. Co-expression with RAMP1 results in CRLR binding to CGRP, whereas association with RAMP2 or 3 results in ADM binding. The ADM receptor, a seven transmembrane G protein-coupled receptor, specifically binds ADM and is highly expressed in heart, brain, skeletal muscle, the immune system, adrenal gland and liver. The ADM receptor mediates the signals produced by ADM through G proteins, which activate adenylate cyclase.
Specifications
| GPR182 | |
| Polyclonal | |
| Unconjugated | |
| GPR182 | |
| 7TMR; ADMR; adrenomedullin receptor; AMR; AM-R; G protein-coupled receptor 182; G10D; G10-D; gamrh; Gpcr17; Gpcr22; GPR182; G-protein coupled receptor 182; HAMR; hrhAMR; MB10; NOW; RAMR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11318 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O15218 | |
| GPR182 | |
| Synthetic 18 amino acid peptide from 2nd extracellular domain of human GPR182 / ADMR. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.