Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR183 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533671
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533671 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533671 Supplier Invitrogen™ Supplier No. PA533671
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%) Bovine (94%) Hamster, Panda, Bat, Dog, Pig (89%) Marmoset, Horse (83%).

GPR183 was identified by the up-regulation of its expression upon Epstein-Barr virus infection of primary B lymphocytes. GPR183 is predicted to encode a G protein-coupled receptor that is most closely related to the thrombin receptor. Expression of this gene was detected in B-lymphocyte cell lines and lymphoid tissues but not in T-lymphocyte cell lines or peripheral blood T lymphocytes. The function of this gene is unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR183
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR183
Gene Accession No. P32249
Gene Alias EBI 2; Ebi2; EBV-induced gene 2; EBV-induced G-protein coupled receptor 2; EBV-induced G-protein coupled receptor 2 homolog; Epstein-Barr virus induced gene 2; Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupled receptor); epstein-Barr virus-induced G-protein coupled receptor 2; Epstein-Barr virus-induced G-protein coupled receptor 2 homolog; G protein-coupled receptor 183; GPR183; G-protein coupled receptor 183; RP11-461N23.2
Gene Symbols GPR183
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from 2nd extracellular domain of human GPR183 / EBI2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1880
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.