Learn More
Invitrogen™ GPR19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114206
Description
Immunogen sequence: LNLLFLLSWL PFHVAQLWHP HEQDYKKSSL VFTAITWISF SSSASKPTLY For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1, 000 members and is the largest known class of molecular targets with proven therapeutic value. GPR19 is an orphan receptor found primarily in the central nervous system. GPR19 was also found in ovary and testis and in human embryonic stem cells. GPR19 has been postulated to play a role in cancer development and increased levels of GPR19 mRNA were found in metastatic melanoma, small cell lung carcinoma and pancreas islet cell carcinoma.
Specifications
GPR19 | |
Polyclonal | |
Unconjugated | |
GPR19 | |
G protein-coupled receptor 19; GPR19; GPR-NGA; Probable G-protein coupled receptor 19 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2842 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q15760 | |
GPR19 | |
Immunogen sequence: LNLLFLLSWL PFHVAQLWHP HEQDYKKSSL VFTAITWISF SSSASKPTLY (aa 281-330). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.