Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533677
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Pig, Bat (100%) Rat, Bovine (93%).
Human follicle stimulating hormone is a glycoprotein produced from urine of post-menopausal women and having a total molecular mass of 35,500 Dalton. HuFSH is a heterodimeric hormone consisting of 165 amino acids a chain (non-active) and 115 amino acids b chain (active). HuFSH induces maturation of Graafian follicles of the ovary, promotes development of germinal cells in males and activates cytosolic tyrosine kinase. The HuFSH is purified by proprietary chromatographic techniques.
Specifications
| GPR21 | |
| Polyclonal | |
| Unconjugated | |
| GPR21 | |
| C230004C13Rik; G protein-coupled receptor 21; G protein-coupled receptor GPR21; Gpr21; Probable G-protein coupled receptor 21 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100155256, 2844 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q99679 | |
| GPR21 | |
| Synthetic 14 amino acid peptide from 3rd transmembrane domain of human GPR21. | |
| 50 μg | |
| Primary | |
| Human, Pig, Bat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction