Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR22 Antibody (455108), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4500Z
Description
GPR22 Monoclonal antibody specifically detects GPR22 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
GPR22 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), CyTOF | |
455108 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
G protein-coupled receptor 22, MGC129847, probable G-protein coupled receptor 22 | |
HEK293 human embryonic kidney cell line transfected with human GPR22, Met1-Asp433, Accession # Q99680 | |
0.1 mL | |
GPCR | |
2845 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction