Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532818
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532818 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532818 Supplier Invitrogen™ Supplier No. PA532818
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Mouse, Rat, Bat, Bovine, Hamster, Elephant, Panda, Horse, Rabbit, Pig, Turkey, Chicken, Lizard, Xenopus, Pufferfish, Zebrafish, Stickleback (100%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1, 000 members and is the largest known class of molecular targets with proven therapeutic value. GPR22 is an orphan G-protein-coupled receptor, belonging to the class A rhodopsin-like GPCRs. GPR22 is highly expressed in several human brain regions, heart and coronary arteries. GPR22 expression is also observed in normal pancreas and in pancreatic, colorectal, prostate and liver cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR22
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR22
Gene Accession No. Q8BZL4, Q99680
Gene Alias 2900068K05Rik; AW061316; G protein-coupled receptor 22; GPR22; G-protein coupled receptor 22; MGC129847; probable G-protein coupled receptor 22; tcag7.108
Gene Symbols GPR22
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminal cytoplasmic domain of human GPR22.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100146673, 100345158, 100519967, 100753963, 2845, 298944, 73010, 769379, 783685
Target Species Human, Mouse, Rat, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Chicken, Frog, Zebrafish, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.