Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | GPR22 |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GPR22 Polyclonal antibody specifically detects GPR22 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
GPR22 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
G protein-coupled receptor 22, MGC129847, probable G-protein coupled receptor 22 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GPR22 (NP_005286.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS, 50% glycerol, pH7.3 | |
2845 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title