Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | GPR22 |
---|---|
Concentration | 0.16 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2618
![]() ![]() |
Proteintech Group Inc
199511AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody recognize the C-term of GPR22.
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1, 000 members and is the largest known class of molecular targets with proven therapeutic value. GPR22 is an orphan G-protein-coupled receptor, belonging to the class A rhodopsin-like GPCRs. GPR22 is highly expressed in several human brain regions, heart and coronary arteries. GPR22 expression is also observed in normal pancreas and in pancreatic, colorectal, prostate and liver cancer.Specifications
GPR22 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8BZL4, Q99680 | |
2845, 298944, 73010 | |
Peptide | |
Primary | |
-20°C | |
GPR22 |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
G protein coupled receptor 22, GPR22 | |
GPR22 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title