Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR26 Antibody (891318), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8449H
Description
GPR26 Monoclonal antibody specifically detects GPR26 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| GPR26 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 891318 | |
| Flow Cytometry, CyTOF-ready | |
| DKFZp761H2121, FLJ33774, G protein-coupled receptor 26, G-protein coupled receptor 26, MGC138216 | |
| NS0 mouse myeloma cell line transfected with human GPR26, Met1-Glu337, Accession # Q8NDV2 | |
| 0.1 mL | |
| GPCR | |
| 2849 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction