Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533684
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Elephant, Panda, Dog, Horse, Pig (100%) Mouse, Rat, Rabbit (94%).
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR26 is a brain-specific orphan GPCR with a significant level of constitutive activity. Its effect is mediated by G(s)-alpha protein that stimulate adenylate cyclase, resulting in an elevation of intracellular cAMP. In the CNS, the highest GPR26 expression is in the amygdala, hippocampus and thalamus. Weak expression is detected in testis. GPR26 is down-regulated in glioblastoma. Depletion of GPR26 has been shown to increase fat storage in C. elegans, whereas GPR26 deficiency in the hypothalamus is associated with high genetic susceptibility to the onset of obesity in mice. GPR26 deficiency also causes metabolic complications commonly associated with obesity, including glucose intolerance, hyperinsulinemia, and dyslipidemia.
Specifications
GPR26 | |
Polyclonal | |
Unconjugated | |
Gpr26 | |
9630036A11Rik; A730021D22; DKFZp761H2121; FLJ33774; G protein-coupled receptor 26; Gpr26; G-protein coupled receptor 26; MGC138216; probable G-protein coupled receptor 26 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100057676, 100154833, 2849, 486926 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8NDV2 | |
Gpr26 | |
Synthetic 17 amino acid peptide from 2nd extracellular domain of human GPR26. | |
50 μg | |
Primary | |
Human, Canine, Monkey, Pig, Horse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction