Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR27 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | GPR27 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
GPR27 Polyclonal specifically detects GPR27 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GPR27 | |
Polyclonal | |
Rabbit | |
GPCR | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
2850 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:NRELRDCFRAQFPCCQSPRTTQATHPCDLKGIGL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
Unconjugated | |
RUO | |
Human | |
G protein-coupled receptor 27, probable G-protein coupled receptor 27, SREB1Super conserved receptor expressed in brain 1 | |
GPR27 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title