Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533685
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533685 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533685 Supplier Invitrogen™ Supplier No. PA533685
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Mouse, Rat, Elephant, Panda (100%); Opossum (89%). For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

G protein coupled receptors (GPCRs) are a large family of receptors that have a similar structure characterized by 7 transmembrane domains. Activation of GPCRs by extracellular stimuli such as neurotransmitters, hormones, or light induces an intracellular signaling cascade mediated by heterotrimeric GTP-binding proteins, or G proteins. The 375 amino acid GPCR GPR27 protein contains several features of the GPCR family, including 7 transmembrane domains, potential N linked glycosylation sites in the N terminal extracellular domain, and conserved cysteine residues, which may form a disulfide bridge, in the first and second extracellular loops.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR27
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR27
Gene Accession No. O54897, Q9JJH3, Q9NS67
Gene Alias G protein-coupled receptor 27; GPR27; Probable G-protein coupled receptor 27; SREB1; Super conserved receptor expressed in brain 1
Gene Symbols GPR27
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human GPR27.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14761, 2850, 65275
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.