Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR27 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533686
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Marmoset (100%) Mouse, Rat, Elephant, Panda (85%).
G protein coupled receptors (GPCRs) are a large family of receptors that have a similar structure characterized by 7 transmembrane domains. Activation of GPCRs by extracellular stimuli such as neurotransmitters, hormones, or light induces an intracellular signaling cascade mediated by heterotrimeric GTP-binding proteins, or G proteins. The 375 amino acid GPCR GPR27 protein contains several features of the GPCR family, including 7 transmembrane domains, potential N linked glycosylation sites in the N terminal extracellular domain, and conserved cysteine residues, which may form a disulfide bridge, in the first and second extracellular loops.
Specifications
GPR27 | |
Polyclonal | |
Unconjugated | |
GPR27 | |
G protein-coupled receptor 27; GPR27; Probable G-protein coupled receptor 27; SREB1; Super conserved receptor expressed in brain 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2850 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9NS67 | |
GPR27 | |
Synthetic 20 amino acid peptide from C-terminus of human GPR27. | |
50 μg | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction