Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533688
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533688 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533688 Supplier Invitrogen™ Supplier No. PA533688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Marmoset, Panda, Pig (100%) Gibbon, Monkey, Bovine, Elephant, Horse (93%) Mouse, Rat, Rabbit (87%).

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1, 000 members and is the largest known class of molecular targets with proven therapeutic value. GPR3 is an orphan receptor with constitutive G signaling activity. GPR3 modulates a number of brain functions, including behavioral responses to stress, neurite outgrowth and amyloid-beta peptide generation suggesting that it may play a role in Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR3
Gene Accession No. P46089
Gene Alias ACCA; ACCA orphan receptor; adenylate cyclase constitutive activator; G protein-coupled receptor 3; G protein-coupled receptor R4; Gpcr20; Gpcr21; Gpcr3; GPR3; G-protein coupled receptor 3; G-protein-coupled receptor R4; Hacca; OTTHUMP00000043209
Gene Symbols GPR3
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from 1st extracellular domain of human GPR3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100512611, 2827
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.