Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR30 Polyclonal Antibody, Bioss
SDP

Catalog No. 502281204
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1204 Supplier Bioss Supplier No. BS20643R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR30 Polyclonal antibody specifically detects GPR30 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen GPR30
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene GPER1
Gene Accession No. O08878, Q8BMP4, Q99527
Gene Alias 6330420K13Rik; CEPR; Ceprl; chemoattractant receptor-like 2; chemokine receptor-like 2; CMKRL2; constitutively expressed peptide-like receptor; constitutively expressed peptide-like receptor like; DRY12; FEG-1; Flow-induced endothelial G-protein coupled receptor 1; G protein-coupled estrogen receptor 1; G protein-coupled receptor 30; GPCR-Br; GPER; GPER1; Gpr30; GPR41; G-protein coupled estrogen receptor 1; G-protein coupled receptor 30; G-protein coupled receptor 41; heptahelix receptor; IL8-related receptor DRY12; LERGU; LERGU2; leucine rich protein in GPR30 3'UTR; LyGPR; Lymphocyte-derived G-protein coupled receptor; membrane estrogen receptor; mER
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPR30
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171104, 2852, 76854
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.