Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533692
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533692 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533692 Supplier Invitrogen™ Supplier No. PA533692
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla (100%).

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR31 is a high-affinity receptor for 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid (12-S-HETE). 12-(S)-HETE is an arachidonic acid metabolite secreted by platelets and tumor cells, and known to induce endothelial cells retraction allowing invasive cell access to the subendothelial matrix, which is a critical step for extravasation or metastasis. Ligand-binding leads to activation of MAPK and NF-kappa-B pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR31
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR31
Gene Accession No. O00270
Gene Alias 12-(S)-HETE acid receptor; 12-(S)-HETE receptor; 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid (HETE) receptor; 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid receptor; 12-HETER; bA517H2.2 (G protein-coupled receptor 31); G protein-coupled receptor 31; GPR31; G-protein coupled receptor 31; HETER; HETER1; hydroxyeicosatetraenoic (HETE) acid receptor 1; probable G-protein coupled receptor 31; TCP10C
Gene Symbols GPR31
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from 2nd extracellular domain of human GPR31.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2853
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.