Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR31 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982949
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2949 Supplier Bioss Supplier No. BS13530R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR31 is a high-affinity receptor for 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid (12-S-HETE). 12-(S)-HETE is an arachidonic acid metabolite secreted by platelets and tumor cells, and known to induce endothelial cells retraction allowing invasive cell access to the subendothelial matrix, which is a critical step for extravasation or metastasis. Ligand-binding leads to activation of MAPK and NF-kappa-B pathways.

Specifications

Antigen GPR31
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GPR31
Gene Accession No. O00270
Gene Alias 12-(S)-HETE acid receptor; 12-(S)-HETE receptor; 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid (HETE) receptor; 12-(S)-hydroxy-5,8,10,14-eicosatetraenoic acid receptor; 12-HETER; bA517H2.2 (G protein-coupled receptor 31); G protein-coupled receptor 31; GPR31; G-protein coupled receptor 31; HETER; HETER1; hydroxyeicosatetraenoic (HETE) acid receptor 1; probable G-protein coupled receptor 31; TCP10C
Gene Symbols GPR31
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human G protein coupled receptor 31.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2853
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.