Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3021 Supplier Thermo Scientific Supplier No. PA3021
Add to Cart
Add to Cart

GPR32 Polyclonal Antibody

GPR32 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR32
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene GPR32
Gene Accession No. O75388
Gene Alias G protein-coupled receptor 32; GPCRW; GPR32; Probable G-protein coupled receptor 32; resolvin D1 receptor; RVDR; RVDR1
Gene Symbols GPR32
Host Species Rabbit
Immunogen KLH conjugated Cys-LARAFGEEEFLSS-Abu-PRGNAPRE.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2854
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.