Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533696
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533696 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533696 Supplier Invitrogen™ Supplier No. PA533696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR32 Polyclonal Antibody for IHC (P)

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. The intronless GPR32 gene encodes an orphan G protein-coupled receptor that binds to resolvin D1 and lipoxin A4. Resolvin D1 binding to GPR32 has been linked to resolution of pulmonary inflammation, suppression of TGFbeta induced epithelial mesenchymal transition of A549 lung cancer cells. Resolvins are being evaluated as new drug candidates acting at the resolution phase of chronic inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR32
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR32
Gene Accession No. O75388
Gene Alias G protein-coupled receptor 32; GPCRW; GPR32; Probable G-protein coupled receptor 32; resolvin D1 receptor; RVDR; RVDR1
Gene Symbols GPR32
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 2nd extracellular domain of human GPR32.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2854
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.