Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR34 Antibody (419859), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4617B
Description
GPR34 Monoclonal antibody specifically detects GPR34 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
GPR34 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
419859 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
G protein-coupled receptor 34, probable G-protein coupled receptor 34 | |
NS0 mouse myeloma cell line transfected with human GPR34, Accession # Q9UPC5 | |
0.1 mL | |
GPCR | |
2857 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction