Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR35 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 5 publications

$206.50 - $512.00

Specifications

Antigen GPR35
Concentration 1.0 mg/mL
Dilution Western Blot 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224640SS
SDP
View Documents
Novus Biologicals
NBP224640SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP224640
SDP
View Documents
Novus Biologicals
NBP224640
0.1 mg
Each for $512.00
Only null left
Add to Cart
 
Description

Description

GPR35 Polyclonal specifically detects GPR35 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

GPR35
Western Blot 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Human, Mouse
G protein-coupled receptor 35, G-protein coupled receptor 35, KYNA receptor, Kynurenic acid receptor
GPR35
IgG
Immunogen affinity purified
1.0 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
GPCR
PBS with 0.05% Sodium Azide
2859
A portion of amino acids 275-309 of human G Protein-Coupled Receptor GPR35 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.