Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR35 Monoclonal Antibody (C3), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543795
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543795 100 μL
PIMA543796 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543795 Supplier Invitrogen™ Supplier No. MA543795
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Acts as a receptor for kynurenic acid, an intermediate in the tryptophan metabolic pathway. The activity of this receptor is mediated by G-proteins that elicit calcium mobilization and inositol phosphate production through G(qi/o) proteins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR35
Applications Western Blot
Classification Monoclonal
Clone C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene GPR35
Gene Accession No. Q9HC97
Gene Alias G protein-coupled receptor 35; Gpr35; G-protein coupled receptor 35; KPG_007; KYNA receptor; kynurenic acid receptor
Gene Symbols GPR35
Host Species Mouse
Immunogen Arg240-Ala309 (Accession Q9HC97), with N-terminal His and GST Tag.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2859
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.