Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR37 Antibody (436923), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4450Y
Description
GPR37 Monoclonal antibody specifically detects GPR37 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
GPR37 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), CyTOF | |
436923 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
EDNRBL, Endothelin B receptor-like protein 1, G protein-coupled receptor 37 (endothelin receptor type B-like), hET(B)R-LP, PAELRETBR-LP-1, Parkin-associated endothelin receptor-like receptor, probable G-protein coupled receptor 37 | |
NS0 mouse myeloma cell line transfected with human GPR37, Ala27-Cys613, Accession # O15354 | |
0.1 mL | |
GPCR | |
2861 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction