Learn More
Invitrogen™ GPR37 Polyclonal Antibody

Description
GPR37 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | GPR37 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | GPR37 |
| Gene Accession No. | O15354, Q9QY42 |
| Gene Alias | AI848630; EDNRBL; endothelin B receptor-like protein 1; ETBR-LP-1; G protein-coupled receptor 37; G protein-coupled receptor 37 (endothelin receptor type B-like); Gpcr/cns1; Gpr37; G-protein coupled receptor 37; G-protein coupled receptor CNS1; hET(B)R-LP; Het-b-r-lp; Pael receptor; PAELR; Pael-R; parkin-associated endothelin B-like receptor; Parkin-associated endothelin receptor-like receptor; probable G-protein coupled receptor 37; prosaposin receptor GPR37 |
| Gene Symbols | GPR37 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.