Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR37L1 Antibody (444532), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4449MFV450
Description
GPR37L1 Monoclonal antibody specifically detects GPR37L1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| GPR37L1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR, Neuroscience, Signal Transduction | |
| 9283 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 444532 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| endothelin B receptor-like protein 2, ET(B)R-LP-2, ETBRLP2, ETBR-LP-2endothelin type b receptor-like protein 2, G protein-coupled receptor 37 like 1, G-protein coupled receptor 37 like 1, G-protein coupled receptor 37-like 1 | |
| NS0 mouse myeloma cell line transfected with human GPR37L1, Accession # O60883 | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction