Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR39 Antibody (189702) [DyLight 488], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3348K
Description
GPR39 Monoclonal antibody specifically detects GPR39 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
GPR39 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
189702 | |
Flow Cytometry, CyTOF-ready | |
G protein-coupled receptor 39, G-protein coupled receptor 39, MGC149541 | |
NS0 mouse myeloma cell line transfected with human GPR39, Met1-Val453, Accession # BAG37409 | |
0.1 mL | |
GPCR, Lipid and Metabolism | |
2863 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction