Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR39 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NLS142F
Description
GPR39 Polyclonal specifically detects GPR39 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GPR39 | |
Polyclonal | |
Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 | |
O43194 | |
GPR39 | |
A synthetic peptide made to an internal region of the human GPR39 protein (within residues 400-500). [Swiss-Prot O43194] | |
0.05 mL | |
GPCR, Lipid and Metabolism | |
2863 | |
Store at 4C in the dark. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
FITC | |
PBS with 0.05% Sodium Azide | |
G protein-coupled receptor 39, G-protein coupled receptor 39, MGC149541 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction