Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR39 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585702
Description
Keep as concentrated solution. Positive Control: U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. G protein-coupled receptor 39 is an orphan receptor in the Ghrelin family pathway. Various studies have shown that GPR39 plays important roles in hypertension, adipogenesis and that overexpression of GPR39 contributes to malignant development of human esophageal squamous cell carcinoma.
Specifications
GPR39 | |
Polyclonal | |
Unconjugated | |
GPR39 | |
4933415E13Rik; G protein-coupled receptor 39; GPR39; G-protein coupled receptor 39 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2863 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.89 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O43194 | |
GPR39 | |
Recombinant protein encompassing a sequence within the C-terminus region of human GPR39. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction